CAS RN 66000-39-3
IUPAC Nome:
1-(4-chlorophenyl)-1H-pyrazol-3-amine
Sinonimi:
1-(4-chlorophenyl)pyrazol-3-amine
Peso molecolare (g/mol):
193.63
Formula molecolare:
C9H8ClN3
InChi Key:
SHCVEWFQFUKSDP-UHFFFAOYSA-N
SMILES:
NC1=NN(C=C1)C1=CC=C(Cl)C=C1