CAS RN 6972-82-3
IUPAC Nome:
5,6-diamino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Sinonimi:
5,6-diamino-1-methyl-3H-pyrimidine-2,4-dione
Peso molecolare (g/mol):
156.15
Formula molecolare:
C5H8N4O2
InChi Key:
PSIJQVXIJHUQPJ-UHFFFAOYSA-N
SMILES:
CN1C(=O)NC(=O)C(N)=C1N
1
–
1
di
1
risultati
1
6-Diamino-1-methyluracil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.