CAS RN 69880-85-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
3-Amino-3-deoxy-D-mannose Hydrochloride, TRC
CAS: 69880-85-9 Formula molecolare: C6 H13 N O5 . Cl H Molecular Weight (g/mol): 215.63 IUPAC Name: (2S,3S,4S,5R)-3-amino-2,4,5,6-tetrahydroxyhexanal;hydrochloride SMILES: Cl.N[C@H]([C@H](O)C=O)[C@H](O)[C@H](O)CO