CAS RN 710-11-2
IUPAC Nome:
2-oxo-4-phenylbutanoic acid
Sinonimi:
2-oxo-4-phenylbutanoic acid
Peso molecolare (g/mol):
178.19
Formula molecolare:
C10H10O3
InChi Key:
PPKAIMDMNWBOKN-UHFFFAOYSA-N
SMILES:
OC(=O)C(=O)CCC1=CC=CC=C1
1
–
1
di
1
risultati
1
2-Oxo-4-phenylbutanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.