CAS RN 859833-20-8
IUPAC Nome:
methyl({[3-(thiophen-2-yl)phenyl]methyl})amine
Sinonimi:
methyl({[3-(thiophen-2-yl)phenyl]methyl})amine
Peso molecolare (g/mol):
203.3
Formula molecolare:
C12H13NS
InChi Key:
BIULJOFRQMLCQK-UHFFFAOYSA-N
SMILES:
CNCC1=CC=CC(=C1)C1=CC=CS1
1
–
1
di
1
risultati
1
N-Methyl-N-(3-thien-2-ylbenzyl)amine, 97%, Thermo Scientific™
CAS: 859833-20-8 Formula molecolare: C12H13NS Molecular Weight (g/mol): 203.30 Numero MDL: MFCD08435869 InChI Key: BIULJOFRQMLCQK-UHFFFAOYSA-N Sinonimo: n-methyl-n-3-thien-2-ylbenzyl amine,methyl 3-thiophen-2-yl phenyl methyl amine,methanamine,n-3-2-thienyl phenyl methylene,n-methyl-1-3-thiophen-2-yl phenyl methanamine,n-methyl-n-3-then-2-ylbenzyl amne,methyl 3-2-thienyl phenyl methyl amine,n-methyl-1-3-thien-2-ylphenyl methylamine PubChem CID: 18525772 SMILES: CNCC1=CC=CC(=C1)C1=CC=CS1