CAS RN 86087-24-3
IUPAC Nome:
oxolan-3-ol
Sinonimi:
3-hydroxytetrahydrofuran
Peso molecolare (g/mol):
88.11
Formula molecolare:
C4H8O2
InChi Key:
XDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILES:
OC1CCOC1
1
–
3
di
3
risultati
3
(R)-(-)-3-Hydroxytetrahydrofuran, 98%, Thermo Scientific™
CAS: 86087-24-3 Formula molecolare: C4H8O2 Molecular Weight (g/mol): 88.106 Numero MDL: MFCD00067101 InChI Key: XDPCNPCKDGQBAN-SCSAIBSYSA-N Sinonimo: r---3-hydroxytetrahydrofuran,r-tetrahydrofuran-3-ol,r-3-hydroxytetrahydrofuran,3r-oxolan-3-ol,3r-tetrahydrofuran-3-ol,r-+-3-hydroxytetrahydrofuran,r-tetrahydro-furan-3-ol,3-furanol, tetrahydro-, 3r,pubchem7071,2-furanyl,tetrahydro-3-hydroxy-, r-9ci PubChem CID: 641512 IUPAC Name: (3R)-oxolan-3-ol SMILES: C1COCC1O