CAS RN 86438-79-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Cocamidopropyl Betaine (~30% aqueous solution), TRC
CAS: 86438-79-1 Formula molecolare: C19H38N2O3 Molecular Weight (g/mol): 342.52 Sinonimo: Cocamidopropyl Betaine IUPAC Name: 2-((3-dodecanamidopropyl)dimethylammonio)acetate SMILES: CCCCCCCCCCCC(NCCC[N+](C)(C)CC([O-])=O)=O