CAS RN 864754-19-8
IUPAC Nome:
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[(thiophen-2-yl)methyl]-1H-pyrazole
Sinonimi:
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(thiophen-2-ylmethyl)pyrazole
Peso molecolare (g/mol):
290.19
Formula molecolare:
C14H19BN2O2S
InChi Key:
ANSBFDYOIUZSNT-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CN(CC2=CC=CS2)N=C1