CAS RN 86482-18-0
IUPAC Nome:
(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid; (2S,5R,6R)-6-[(2R)-2-carboxy-2-(thiophen-3-yl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Sinonimi:
ticarcillin clavulanate
Peso molecolare (g/mol):
583.58
Formula molecolare:
C23H25N3O11S2
InChi Key:
XWMVMWTVLSLJGY-FAJPTIRJSA-N
SMILES:
OC\C=C1/O[C@@H]2CC(=O)N2[C@H]1C(O)=O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C(O)=O)C3=CSC=C3)C(=O)N2[C@H]1C(O)=O