CAS RN 874819-71-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
(2-Nitrophenyl)phosphoric Triamide, TRC
CAS: 874819-71-3 Formula molecolare: C6 H9 N4 O3 P Molecular Weight (g/mol): 216.13 Sinonimo: Phosphoric triamide, (2-nitrophenyl)- (9CI),N-(2-Nitrophenyl)phosphoric triamide,N-(2-Nitrophenyl)phosphoric acid triamide,N-(2-Nitrophenyl)phosphoric Triamide IUPAC Name: N-diaminophosphoryl-2-nitroaniline SMILES: NP(=O)(N)Nc1ccccc1[N+](=O)[O-]