CAS RN 875755-24-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
R&D Systems™ NS 8593 Hydrochloride
A Tocris product, part of R&D Systems. Selective negative modulator of KCa2 (SK) channels; inhibits SK currents
2
Tocris Bioscience™ NS 8593 Hydrochloride
Selective negative modulator of KCa2 (SK) channels; inhibits SK currents
3
Tocris Bioscience™ NS 8593 Hydrochloride
Selective negative modulator of KCa2 (SK) channels; inhibits SK currents