CAS RN 877-37-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2-Bromo-1-(4-chlorophenyl)propan-1-one, TRC
CAS: 877-37-2 Formula molecolare: C9H8BrClO Molecular Weight (g/mol): 247.52 IUPAC Name: 2-bromo-1-(4-chlorophenyl)propan-1-one SMILES: CC(Br)C(=O)c1ccc(Cl)cc1