CAS RN 877877-35-5
IUPAC Nome:
N-[4-(2-tert-butylbenzenesulfonyl)phenyl]-2,3,4-trihydroxy-5-{[2-(propan-2-yl)phenyl]methyl}benzamide
Sinonimi:
N-[4-(2-tert-butylbenzenesulfonyl)phenyl]-2,3,4-trihydroxy-5-[(2-isopropylphenyl)methyl]benzamide
Peso molecolare (g/mol):
573.7
Formula molecolare:
C33H35NO6S
InChi Key:
PQAPVTKIEGUPRN-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1CC1=C(O)C(O)=C(O)C(=C1)C(=O)NC1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1C(C)(C)C
1
–
1
di
1
risultati
1
TW-37, MedChemExpress
MedChemExpress TW-37 is a potent Bcl-2 inhibitor with Ki values of 260, 290 and 1110 nM for Mcl-1, Bcl-2 and Bcl-xL, respectively.