CAS RN 87864-14-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
2-Chloro-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-Chloro-N-[2-(diethylamino)ethyl]-4-quinolinecarboxamide, TRC
CAS: 87864-14-0 Formula molecolare: C16 H20 Cl N3 O Molecular Weight (g/mol): 305.8 Sinonimo: 4-Quinolinecarboxamide, 2-chloro-N-[2-(diethylamino)ethyl]-,Cinchoninamide, 2-chloro-N-(2-diethylaminoethyl)- (6CI) IUPAC Name: 2-chloro-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide SMILES: CCN(CC)CCNC(=O)c1cc(Cl)nc2ccccc12