CAS RN 879896-49-8
IUPAC Nome:
[1-methyl-3-(thiophen-2-yl)-1H-pyrazol-5-yl]methanol
Sinonimi:
[2-methyl-5-(thiophen-2-yl)pyrazol-3-yl]methanol
Peso molecolare (g/mol):
194.25
Formula molecolare:
C9H10N2OS
InChi Key:
CHKYTDUITSTBNJ-UHFFFAOYSA-N
SMILES:
CN1N=C(C=C1CO)C1=CC=CS1