CAS RN 880155-70-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
CNT2 inhibitor-1, MedChemExpress
MedChemExpress CNT2 inhibitor-1 is a potent concentrative nucleoside transporter 2 Inhibitor (CNT2), with an IC50 of 640 nM for hCNT2.