CAS RN 88057-03-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
(R)-Coniine Hydrochloride (~25% S-isomer), TRC
CAS: 88057-03-8 Formula molecolare: C8 H17 N . H Cl Molecular Weight (g/mol): 163.69 Sinonimo: (R)-2-Propylpiperidine Hydrochloride,(-)-Coniine hydrochloride IUPAC Name: (2R)-2-propylpiperidine;hydrochloride SMILES: Cl.CCC[C@@H]1CCCCN1