CAS RN 881-57-2
IUPAC Nome:
5-formyl-2-methoxyphenyl acetate
Sinonimi:
5-formyl-2-methoxyphenyl acetate
Peso molecolare (g/mol):
194.19
Formula molecolare:
C10H10O4
InChi Key:
ZVPGTXJXZIXWGR-UHFFFAOYSA-N
SMILES:
COC1=C(OC(C)=O)C=C(C=O)C=C1
1
–
3
di
3
risultati
1
O-Acetyl Isovanillin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
O-Acetyl Isovanillin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
O-Acetyl Isovanillin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.