CAS RN 882405-89-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
BBT594, MedChemExpress
MedChemExpress BBT594 is a potent receptor tyrosine kinase RET inhibitor, used for cancer treatment.