CAS RN 942-58-5
CAS RN 942-58-5
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Allyl O-Cumenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Allyl O-Cumenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Allyl O-Cumenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.