CAS RN 94367-21-2
CAS RN 94367-21-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
MedChemExpress Suc-Leu-Leu-Val-Tyr-AMC
MedChemExpress Suc-Leu-Leu-Val-Tyr-AMC is a fluorogenic substrate.