CAS RN 943858-40-0
CAS RN 943858-40-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
(BetaR)-Beta-[[(S)-(4-Methylphenyl)sulfinyl]amino]-4-[[tris(1-methylethyl)silyl]oxy]benzenepropanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(BetaR)-Beta-[[(S)-(4-Methylphenyl)sulfinyl]amino]-4-[[tris(1-methylethyl)silyl]oxy]benzenepropanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(BetaR)-Beta-[[(S)-(4-Methylphenyl)sulfinyl]amino]-4-[[tris(1-methylethyl)silyl]oxy]benzenepropanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.