CAS RN 944153-47-3
CAS RN 944153-47-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
SMER18, MedChemExpress
MedChemExpress SMER18 is a small molecule enhancer of rapamycin which act as a mTOR-independent autophagy inducer.