CAS RN 94535-50-9
CAS RN 94535-50-9
IUPAC Nome:
(3S,4R)-3-hydroxy-2,2-dimethyl-4-(2-oxopyrrolidin-1-yl)-3,4-dihydro-2H-1-benzopyran-6-carbonitrile
Sinonimi:
(-)-cromakalim lemakalim levcromakalim levcromakelim
Peso molecolare (g/mol):
286.33
Formula molecolare:
C16H18N2O3
InChi Key:
TVZCRIROJQEVOT-CABCVRRESA-N
SMILES:
CC1(C)OC2=CC=C(C=C2[C@H]([C@@H]1O)N1CCCC1=O)C#N
1
–
2
di
2
risultati
1
Promozioni disponibili
Tocris Bioscience™ Levcromakalim
Tocris Bioscience™ Levcromakalim
CAS: 94535-50-9 Formula molecolare: C16H18N2O3
2
Levcromakalim, MedChemExpress
MedChemExpress Levcromakalim ((-)-Cromakalim) is an ATP-sensitive K+ channel (KATP) activator.