CAS RN 946518-61-2
CAS RN 946518-61-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
LY2409881, MedChemExpress
MedChemExpress LY2409881 is a selective IκB kinase β (IKK2) inhibitor with an IC50 of 30 nM.