CAS RN 94801-02-2
CAS RN 94801-02-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
5-Azido-5-deoxy-D-fructose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Azido-5-deoxy-D-fructose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Azido-5-deoxy-D-fructose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.