CAS RN 95-44-3
CAS RN 95-44-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
L-Threose (~0.2 M in Water), TRC
CAS: 95-44-3 Formula molecolare: C4H8O4 Molecular Weight (g/mol): 120.1 Sinonimo: (2R,3S)-2,3,4-Trihydroxy-butanal,[R-(R*,S*)]-2,3,4-Trihydroxy-butanal,L-Threofuranose IUPAC Name: (2R,3S)-2,3,4-trihydroxybutanal SMILES: OC[C@H](O)[C@@H](O)C=O