CAS RN 952718-75-1
CAS RN 952718-75-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
Desmethyl-6,8,10-triene Dutasteride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Desmethyl-6,8,10-triene Dutasteride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.