CAS RN 956136-95-1
CAS RN 956136-95-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Pradigastat, MedChemExpress
MedChemExpress Pradigastat (LCQ-908) is a potent, selective and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor. Pradigastat has anti-obesity and anti-diabetic effects.