CAS RN 957118-49-9
CAS RN 957118-49-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
MK-3207, MedChemExpress
MedChemExpress MK-3207 is a potent and orally bioavailable CGRP receptor antagonist (IC50= 0.12 nM; Ki= 0.024 nM); highly selective versus human AM1, AM2, CTR, and AMY3.