CAS RN 95926-99-1
CAS RN 95926-99-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
2-Methyl-propanoic-3,3,3-d3 Acid, TRC
CAS: 95926-99-1 Formula molecolare: C4 D3 H5 O2 Molecular Weight (g/mol): 91.124 Sinonimo: Propanoic-3,3,3-d3 acid, 2-methyl- (9CI),2-Methylpropanoic-3,3,3-d3 acid,Isobutyric-3,3,3-d6 acid,2-Methylpropanoic Acid-3,3,3-D3,Isobutyric acid D3 (methyl-D3),Isobutyric acid-d3,2-Methyl-d3-propionic Acid IUPAC Name: 3,3,3-trideuterio-2-methylpropanoic acid SMILES: [2H]C([2H])([2H])C(C)C(=O)O
2
2-Methyl-d3-propionic Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.