CAS RN 96258-91-2
CAS RN 96258-91-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.