CAS RN 96512-76-4
CAS RN 96512-76-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
O-Methyl Chlorthalidone, TRC
CAS: 96512-76-4 Formula molecolare: C15 H13 Cl N2 O4 S Molecular Weight (g/mol): 352.793 Sinonimo: 2-Chloro-5-(2,3-dihydro-1-methoxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide,2-Chloro-5-(1-methoxy-3-oxo-1-isoindolinyl)benzenesulfonamide IUPAC Name: 2-chloro-5-(1-methoxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide SMILES: COC1(NC(=O)c2ccccc12)c3ccc(Cl)c(c3)S(=O)(=O)N