CAS RN 96829-59-3
CAS RN 96829-59-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Lipstatin, TRC
CAS: 96829-59-3 Formula molecolare: C29H49NO5 Molecular Weight (g/mol): 491.7 Sinonimo: N-Formyl-L-leucine (1S,3Z,6Z)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]-3,6- dodecadien-1-yl Ester,N-Formyl-L-leucine [2S-[2α(1R*,3Z,6Z),3β]]-1- [(3-Hexyl-4-oxo-2-oxetanyl)methyl]-3,6-dodecadienyl Ester,(-)-Lipstatin SMILES: CCCCCC[C@H]1[C@@H](OC1=O)C[C@H](C/C=C\C/C=C\CCCCC)OC(=O)[C@H](CC(C)C)NC=O