CAS RN 97028-91-6
CAS RN 97028-91-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
4,4-Diethoxy-N,N-diethyl-1-butanamine, TRC
CAS: 97028-91-6 Formula molecolare: C12H27NO2 Molecular Weight (g/mol): 217.35 Sinonimo: 4-(Diethylamino)butanal Diethyl Acetal IUPAC Name: 4,4-diethoxy-N,N-diethylbutan-1-amine SMILES: CCOC(CCCN(CC)CC)OCC