CAS RN 97203-04-8
CAS RN 97203-04-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
4
di
4
risultati
1
7-Dehydroxy Buprenorphine (Buprenorphine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Dehydroxy Buprenorphine (Buprenorphine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
17-(Cyclopropylmethyl)-4,5α-epoxy-6-methoxy-7α-[1-(1,1-dimethylethyl)ethenyl]-6α,14-ethano-14α-morphinan-3-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
7-Dehydroxy Buprenorphine (Buprenorphine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.