CAS RN 977-32-2
CAS RN 977-32-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
4
di
4
risultati
1
3-Oxoandrosta-1,4-dien-17beta-yl Propanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Boldenone Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Boldenone Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Boldenone Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.