CAS RN 98293-79-9
CAS RN 98293-79-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
3-Cyano-6-dimethoxymethyl-5-(4-pyridyl)-2-pyridone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Cyano-6-dimethoxymethyl-5-(4-pyridyl)-2-pyridone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Cyano-6-dimethoxymethyl-5-(4-pyridyl)-2-pyridone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.