CAS RN 98454-50-3
CAS RN 98454-50-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
trans-5-(4-Chlorobutyl)-1-[4-(phenylmethoxy)cyclohexyl]-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans-5-(4-Chlorobutyl)-1-[4-(phenylmethoxy)cyclohexyl]-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
trans-5-(4-Chlorobutyl)-1-[4-(phenylmethoxy)cyclohexyl]-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.