CAS RN 98523-85-4
CAS RN 98523-85-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
17-Hydroxy-3,20-dioxopregna-1,4-diene-11beta,21-diyl Diacetate (Prednisolone 11,21-Diacetate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Prednisolone 11,21-Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.