CAS RN 98632-91-8
CAS RN 98632-91-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
N-Carbobenzyloxy-D-serine-Beta-lactone, TRC
CAS: 98632-91-8 Formula molecolare: C11 H11 N O4 Molecular Weight (g/mol): 221.21 Sinonimo: Carbamic acid, (2-oxo-3-oxetanyl)-, phenylmethyl ester, (R)-,Carbamic acid, [(3R)-2-oxo-3-oxetanyl]-, phenylmethyl ester (9CI),Phenylmethyl N-[(3R)-2-oxo-3-oxetanyl]carbamate,(R)-Benzyl 2-oxooxetan-3-ylcarbamate IUPAC Name: benzyl N-[(3R)-2-oxooxetan-3-yl]carbamate SMILES: O=C(N[C@@H]1COC1=O)OCc2ccccc2