CAS RN 99-11-6
CAS RN 99-11-6
IUPAC Nome:
6-hydroxy-2-oxo-1,2-dihydropyridine-4-carboxylic acid
Sinonimi:
citrazinic acid
Peso molecolare (g/mol):
155.11
Formula molecolare:
C6H5NO4
InChi Key:
CSGQJHQYWJLPKY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=O)NC(O)=C1
1
–
2
di
2
risultati
2
2,6-dihydroxyisonicotinic acid, Thermo Scientific™
CAS: 99-11-6 Formula molecolare: C6H5NO4 Molecular Weight (g/mol): 155.11 Numero MDL: MFCD00006274 InChI Key: CSGQJHQYWJLPKY-UHFFFAOYSA-N Sinonimo: citrazinic acid,2,6-dihydroxyisonicotinic acid,2,6-dihydroxypyridine-4-carboxylic acid,2,6-dihydroxy-4-carboxypyridine,2,6-dihydroxy-4-pyridinecarboxylic acid,kyselina citrazinova,2,6-dihydroxyisonicotinate,4-pyridinecarboxylic acid, 2,6-dihydroxy,citrazinicacid,4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo PubChem CID: 7425 IUPAC Name: 2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid SMILES: OC(=O)C1=CC(=O)NC(O)=C1