CAS RN 99102-04-2
CAS RN 99102-04-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
rac erythro-Dihydro Bupropion, TRC
CAS: 99102-04-2 Formula molecolare: C13 H20 Cl N O Molecular Weight (g/mol): 241.76 Sinonimo: Benzenemethanol, 3-chloro-α-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]-, (αS)-rel- (9CI, ACI),Benzenemethanol, 3-chloro-α-[1-[(1,1-dimethylethyl)amino]ethyl]-, (R*,S*)- (ZCI),rel-(αS)-3-Chloro-α-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]benzenemethanol (ACI),Erythrohydrobupropion,Erythrohydroxybupropion,rel-(1R,2S)-2-(tert-Butylamino)-1-(3-chlorophenyl)-1-propanol,rac erythro-Dihydro Bupropion,(±)-erythro-Dihydrobupropion IUPAC Name: (1R,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-ol SMILES: C[C@H](NC(C)(C)C)[C@H](O)c1cccc(Cl)c1