CAS RN 99159-87-2
CAS RN 99159-87-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
(2R)-3-Hydroxyisovaleroyl Carnitine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-3-Hydroxyisovaleroyl Carnitine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.