CAS RN 99199-60-7
CAS RN 99199-60-7
IUPAC Nome:
6-fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Sinonimi:
6-fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Peso molecolare (g/mol):
196.18
Formula molecolare:
C10H9FO3
InChi Key:
ZNJANLXCXMVFFI-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCC2=C(O1)C=CC(F)=C2
1
–
2
di
2
risultati
1
rac-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.