CAS RN 99326-34-8
CAS RN 99326-34-8
IUPAC Nome:
bis(cycloocta-1,5-diene) rhodium trifluoromethanesulfonate
Sinonimi:
bis(1,5-cyclooctadiene) rhodium triflate
Peso molecolare (g/mol):
468.34
Formula molecolare:
C17H24F3O3RhS
InChi Key:
VUTUHLLWFPRWMT-UHFFFAOYSA-M
SMILES:
[Rh].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1
1
–
1
di
1
risultati
1
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
CAS: 99326-34-8 Formula molecolare: C17H24F3O3RhS Molecular Weight (g/mol): 468.34 Numero MDL: MFCD00143752 InChI Key: GOPBTCMAUMSOBX-QMDOQEJBSA-M Sinonimo: bis 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,bis 1,5-cyclooctadiene rhodium i triflate,rh cod 2 otf,bis 1,5-cyclooctadiene rhodium i trifl,bis 1,5-cyclooctadiene rhodium triflate,rhodium 1+ ion bis 1,5-cyclooctadiene triflate,bis 1,5-cyclooctadiene rhodium i trifluoromethane,n-tert-butyloxycarbonyl-n'-benzyloxymethyl-l-histidine,bis 1,5-cyclooctadiene-trifluoromethanesulfitorhodium i PubChem CID: 5702638 SMILES: [Rh+].[O-]S(=O)(=O)C(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1