CAS RN 99685-96-8
Fullerene powder, 99.5% C{60}
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, hydroxylated, C{60}(OH)∣n,
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD06202029 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene C60, 99.9%
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, sublimed, 99.9+% C{60}
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, 99% C{6}{0}
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, 99.9+% C{60}
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene C60, TRC
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.64 Sinonimo: Buckminsterfullerene,Buckminsterfullerene (C60),Buckyball,C60 Fullerene,Carbon (C60),Carbon (C60) fullerene,Carbon (C60) mol.,Carbon cluster (C60),Carbon, mol. (C60),Follene-60,Footballene,Footballene (C60),Fullerene,Fullerene C60 cluster,Fullerene-60,Fullerene-C60,Icosahedral C60,Nano-C60,Nanom Purple,Nanom Purple N 60S,Nanom Purple ST,Nanom Purple ST-A,Nanom Purple SU,Soccerballene,Tris-PC61BOE,XFC 01,[5,6]Fullerene C60,[60]Fullerene SMILES: c12c3c4c5c6c7c8c9c%10c%11c%12c9c%13c%14c%15c%12c%16c%17c%11c%18c%19c%10c%20c8c6c%21c%22c%20c%19c%23c%24c%18c%17c%25c%26c%16c%15c%27c%28c%14c(c7%13)c5c3c%28c%29c%27c%26c%30c%25c%24c%31c%23c%22c(c4%21)c1c%31c%30c2%29
Fullerene powder, mixed, typically 98% C{60}, 2% C{70}
CAS: 99685-96-8 Formula molecolare: C60 Molecular Weight (g/mol): 720.66 Numero MDL: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Sinonimo: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41