CAS RN 99799-09-4
CAS RN 99799-09-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2-(4-Hydroxycyclohexyl)acetic Acid, TRC
CAS: 99799-09-4 Formula molecolare: C8H14O3 Molecular Weight (g/mol): 158.2 Sinonimo: 4-Hydroxycyclohexaneacetic Acid,(4-Hydroxy-cyclohexyl)-acetic Acid IUPAC Name: 2-(4-hydroxycyclohexyl)acetic acid SMILES: OC1CCC(CC(=O)O)CC1