CAS RN 99833-87-1
CAS RN 99833-87-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
3,4-Dihydro-7-methoxy-2H-1-naphthalenone-O-tosyloxime, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,4-Dihydro-7-methoxy-2H-1-naphthalenone-O-tosyloxime, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,4-Dihydro-7-methoxy-2H-1-naphthalenone-O-tosyloxime, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.