Real Time PCR Master Mixes
Real-time PCR, or qPCR, permits the detection and quantification of target nucleic acids. PCR master mixes can be used to reduce the risk of contamination and other errors related to mixing and pipetting.
Useful Links
Save Now - Exclusive Deals
Product Code 4527036
Product Code 4527035
Complete Your Order - Great Deals
Product Code 7184431
Product Code 7184433
Must Have
Product Code 8015682
FAQ
A master mix for real-time PCR is a pre-prepared solution containing most of the components needed for the PCR reaction, such as DNA polymerase, dNTPs, MgCl2, buffer, and sometimes fluorescent dyes, to ensure consistency and efficiency in the amplification process.
The components of a master mix for RT-PCR include:
- DNA Polymerase: An enzyme to synthesize new DNA strands.
- dNTPs (Deoxynucleotide triphosphates): Building blocks for new DNA synthesis
- MgCl2 (Magnesium chloride): A cofactor required for DNA polymerase activity
- Buffer: Maintains the optimal pH and salt conditions for the reaction
- Fluorescent Dye (e.g., SYBR Green or TaqMan probes): For real-time detection of DNA amplification
- RNase Inhibitor: Prevents degradation of RNA (if reverse transcription is included)
- Reverse Transcriptase: Enzyme to synthesize cDNA from RNA (for RT-PCR)
- Primers: Short DNA sequences that initiate DNA synthesis
These components are combined in specific proportions to ensure efficient and accurate target DNA/RNA amplification.
When buying RT-PCR master mixes, consider the following five factors:
- Ensure the master mix is compatible with your specific real-time PCR instrument and applications.
- Look for mixes that offer high sensitivity and specificity for detecting low-abundance targets and distinguishing between similar sequences.
- Check the stability and shelf life to ensure the mix remains effective over time and under storage conditions.
- Prefer mixes that are ready-to-use and minimize preparation steps, reducing the chance of errors.
- Balance the cost with performance features, considering your budget and the scale of your experiments.